VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Supplier: VWR Chemicals
VWRC352432CEA 166 SGD
VWRC352432C
VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Stains Microscopy and staining reagents
Decalcifier according to Kristenson.
  • pH (25°C) 2.20 --> 2.70