VWR® Media Supplements for Dehydrated Culture Media
Supplier: VWR Chemicals
VWRC602730ZLEA 270 SGD
VWRC602730ZL VWRC968130NL VWRC968290NL VWRC928010NL VWRC928020NL VWRC928030NL VWRC928090NL VWRC928110OL VWRC928140NL VWRC928150NL VWRC928170OL VWRC928180NL VWRC928190NL VWRC928200NL VWRC928230NL VWRC928240OL VWRC928250NL VWRC928260NL VWRC928280NL VWRC928320NL VWRC928330NL VWRC928350NL VWRC928360NL VWRC928370NL VWRC928380NL VWRC928390NL VWRC928300NL VWRC928340NL VWRC303570ZK VWRC335032ZF VWRC928460NL VWRC928080NL VWRC928240NL VWRC355070ZF VWRC84721.0002
VWR® Media Supplements for Dehydrated Culture Media
Media for Microbiology Additives and Supplements for Microbiology
These standard supplements can be used with several culture media to prepare the complete culture media for your specific purpose. Most of these supplements are available as freeze-dried in vials and this ensures.
  • No toxic agent aerosols released upon opening
  • Standard volumes make preparation easier
  • User friendly screw caps
  • Can be stored at room temperature
Order Now
SPECIFICATIONS
Environmentally Preferable