Paramat, Solidification point: 55 - 58 °C, in pastille form, Gurr™, without DMSO

Supplier: VWR Chemicals

VWRC361147BEA 368 SGD
VWRC361147B VWRC361148C
Paramat, Solidification point: 55 - 58 °C, in pastille form, Gurr™, without DMSO
Paraffin wax blended with synthetic polymers

Paramat is a finely filtered blend of specially selected high purity paraffin wax with an addition of polymer to give improved embedding, tissue support and reliable sectioning. Residual moisture in the tissue and traces of intermedium are dispelled. The pastille shape makes for easy handling.


  • Paraffin wax blended with synthetic polymers
  • Solidification point 55 - 58 ° C


Available also with DMSO - see Paramat Extra (VWRC36133)

CAS Number: 8002-74-2

Order Now

Specification Test Results

Cloud point (65 °C) Clear
Cutting/ribboning Complies
Congealing temperature 55 - 58 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR