VWR® Scholten's Culture Medium for Detection of Somatic Coliphages
Supplier: VWR Chemicals
VWRC406103ZAEA 191 SGD
VWRC406103ZA VWRC376234ZA
VWR® Scholten's Culture Medium for Detection of Somatic Coliphages
Media for Microbiology Prepared Media for Microbiology
The Scholten’s Modified broth is described in ISO 10705-2: 2000 - Water Quality - Detection and enumeration of bacteriophages - somatic coliphages. It is used as a growth culture medium for the production and maintenance of the host-bacteria for the counting of bacteriophages.
  • Wide choice of dehydrated or ready-to-use products
  • Room temperature storage for all the products
  • Long shelf life for plates (7 months) and bottles (12 months)
Certifications: In compliance with ISO 10705-2: 2000 for Water Quality.
Order Now
SPECIFICATIONS
Environmentally Preferable