VWR®, Penicillin-streptomycin solution (100X)

Supplier: VWR Chemicals

VWRC392-0406EA 37.1 SGD
VWRC392-0406
VWR®, Penicillin-streptomycin solution (100X)
Cell Culture Media

VWR® penicillin-streptomycin solution (100X) is a large spectrum antibiotic designed to control and destroy gram-negative and gram-positive bacteria.


  • With 10000 units penicillin and 10 mg/ml streptomycin in 0,9% NaCl
  • Sterile: 0,1 μm filtered
  • Use in cell culture applications at 10 ml/L for tissue culture media containing serum
  • Endotoxin level at 10 ml/L of solution: <0,1 EU/ml


Antibiotics, combined with good sterile technique of manipulation of cell culture, help prevent microbiological contamination.

Order Now

SPECIFICATIONS

Environmentally Preferable
Sterility Sterile

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR