VWR®, Cell culture media, Ham's F12
Supplier: VWR Chemicals
VWRC392-0418EA 36.1 SGD
VWRC392-0418
VWR®, Cell culture media, Ham's F12
Cell Culture Media
Ham's F12 was developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells. It is frequently used with supplements for serum-free cultures to support the growth of cells of rodent origin, or as a cloning medium for the culture of myeloma and hybrid cells.
Order Now
SPECIFICATIONS
Environmentally Preferable
Media Base Ham's F12 Nutrient Mixture
Calcium Yes
HEPES No
L-Glutamine Yes
Magnesium Yes
Phenol red Yes
Sodium bicarbonate Yes
Sodium pyruvate Yes
Stable glutamine No