VWR®, Cell culture media, Schneider's Drosophila medium
Supplier: VWR Chemicals
VWRC392-0419EA 81.2 SGD
VWRC392-0419
VWR®, Cell culture media, Schneider's Drosophila medium
Cell Culture Media
Schneider's Drosophila medium promotes rapid growth of both primary and existing cell cultures derived from Drosophila spp and several other dipterans when supplemented with 5 to 20% heat-inactivated foetal bovine serum.