VWR®, Cell culture media, Medium 199
Supplier: VWR Chemicals
VWRC392-0421EA 83 SGD
VWRC392-0421
VWR®, Cell culture media, Medium 199
Cell Culture Media
Medium 199 was developed specifically for nutritional research of chicken fibroblasts. Today, 199 media are widely used for the maintenance of non transformed cells, vaccine and virus production and primary explants of epithelial cells. The media formulated with the Hanks salts are buffered with saline solutions designed for balancing in ambient conditions, and their use in a CO₂ incubator results in a rapid drop in pH of the culture medium.
Order Now
SPECIFICATIONS
Environmentally Preferable
Media Base Medium 199 (M199) Medium
Calcium Yes
HEPES No
L-Glutamine Yes
Magnesium Yes
Phenol red Yes
Sodium bicarbonate Yes
Sodium pyruvate No
Stable glutamine No