VWR®, L-Glutamine
Supplier: VWR Chemicals
VWRC392-0441EA 33.4 SGD
VWRC392-0441
VWR®, L-Glutamine
Cell Culture Media
L-Glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture.
Order Now
SPECIFICATIONS
Environmentally Preferable