Water Nuclease_free

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

VWRC436912CEA 113 SGD
VWRC436912C
Water Nuclease_free
Water

Water, nuclease-free, molecular biology grade (RNase, DNase and protease tested).

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Osmolality (mOsm/kg) Max. 3
DNases Not detected
RNases Not detected
Proteases Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR