Sodium dodecyl sulphate (SDS) ≥99.0% for biochemistry, specially purified

Supplier: VWR Chemicals
Danger

VWRC44215HNEA 443 SGD
VWRC44215HN VWRC44215LT
Sodium dodecyl sulphate (SDS) ≥99.0% for biochemistry, specially purified
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1,1 g/cm³ (20 °C)
Flash Pt: 100 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.0 %
Assay (C12 compounds)(calc.on anhydrous) Min. 98.5 %
IR Spectrum Passes test
Cl (Chloride) Max. 0.03 %
PO4 (Phosphate) Max. 1 ppm
Pb (Lead) Max. 2 ppm
Absorbance (220-350 nm) (3 %) Max. 0.1

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR