VWR® Phosphate Buffered Saline (PBS), High Phosphate
Supplier: VWR Chemicals
VWRC444037EEA 675 SGD
VWRC444037E
VWR® Phosphate Buffered Saline (PBS), High Phosphate
Buffers Electrophoresis Buffers Washing Buffers
Irradiated solution, pH 7,2, for washing blots.
  • NaCl 8,77 g/L
  • Na₂HPO₄, 12H₂0 19,2 g/L
  • NaH₂PO₄, 2H₂O 6,55 g/L