Water, Electran Molecular_Biology

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

VWRC445847DEA 100160 SGD
VWRC445847D
Water, Electran Molecular_Biology
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Osmolality (mOsm/kg) Max. 3
DNases Not detected
RNases Not detected
Proteases Not detected
Fe (Iron) Max. 0.006 ppm
Mg (Magnesium) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR