Nitric acid 69%, ARISTAR® for trace analysis

Supplier: VWR Chemicals
Danger

Synonyms: Nitric acid solution

VWRC450041MEA 213 SGD
VWRC450041M VWRC450042N
Nitric acid 69%, ARISTAR® for trace analysis
Nitric acid
Formula: HNO₃
Boiling Pt: 120,5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 68.5 - 69.5 %
Cl (Chloride) Max. 0.05 ppm
PO4 (Phosphate) Max. 0.1 ppm
Si (as silicate) Max. 0.1 ppm
SO4 (Sulphate) Max. 0.5 ppm
As + Sb (as As) Max. 0.005 ppm
Ag (Silver) Max. 0.005 ppm
Al (Aluminium) Max. 0.020 ppm
Au (Gold) Max. 0.005 ppm
B (Boron) Max. 0.005 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.005 ppm
Bi (Bismuth) Max. 0.005 ppm
Ca (Calcium) Max. 0.05 ppm
Cd (Cadmium) Max. 0.005 ppm
Co (Cobalt) Max. 0.005 ppm
Cr (Chromium) Max. 0.010 ppm
Cu (Copper) Max. 0.005 ppm
Fe (Iron) Max. 0.020 ppm
Ga (Gallium) Max. 0.020 ppm
Ge (Germanium) Max. 0.010 ppm
Hg (Mercury) Max. 0.001 ppm
In (Indium) Max. 0.010 ppm
K (Potassium) Max. 0.010 ppm
Li (Lithium) Max. 0.005 ppm
Mg (Magnesium) Max. 0.010 ppm
Mn (Manganese) Max. 0.005 ppm
Mo (Molybdenum) Max. 0.005 ppm
Na (Sodium) Max. 0.050 ppm
Ni (Nickel) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm
Pt (Platinum) Max. 0.005 ppm
Sn (Tin) Max. 0.020 ppm
Sr (Strontium) Max. 0.005 ppm
Ti (Titanium) Max. 0.005 ppm
Tl (Thallium) Max. 0.005 ppm
V (Vanadium) Max. 0.005 ppm
Zn (Zinc) Max. 0.010 ppm
Zr (Zirconium) Max. 0.005 ppm
Ignition residue (SO4) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR