Formic acid 98,0, ARISTAR® Trace_analysis

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

VWRC450122MEA 884 SGD
VWRC450122M
Formic acid 98,0, ARISTAR® Trace_analysis
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8,2…8,4 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3) II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) Min. 98.0 %
Colour Max. 10 APHA
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 0.5 ppm
Acetic acid Max. 500 ppm
Ag (Silver) Max. 0.001 ppm
Al (Aluminium) Max. 0.005 ppm
Au (Gold) Max. 0.005 ppm
Ba (Barium) Max. 0.002 ppm
Be (Beryllium) Max. 0.001 ppm
Bi (Bismuth) Max. 0.002 ppm
Ca (Calcium) Max. 0.05 ppm
Cd (Cadmium) Max. 0.001 ppm
Co (Cobalt) Max. 0.001 ppm
Cr (Chromium) Max. 0.001 ppm
Cu (Copper) Max. 0.002 ppm
Fe (Iron) Max. 0.010 ppm
Ga (Gallium) Max. 0.005 ppm
Hg (Mercury) Max. 0.005 ppm
In (Indium) Max. 0.002 ppm
K (Potassium) Max. 0.02 ppm
Li (Lithium) Max. 0.001 ppm
Mg (Magnesium) Max. 0.01 ppm
Mn (Manganese) Max. 0.001 ppm
Mo (Molybdenum) Max. 0.001 ppm
Na (Sodium) Max. 0.02 ppm
Ni (Nickel) Max. 0.002 ppm
Pb (Lead) Max. 0.002 ppm
Sn (Tin) Max. 0.001 ppm
Sr (Strontium) Max. 0.001 ppm
Ti (Titanium) Max. 0.001 ppm
Tl (Thallium) Max. 0.001 ppm
V (Vanadium) Max. 0.001 ppm
Zn (Zinc) Max. 0.005 ppm
Non-volatile matter Max. 2 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR