VWR® ARISTAR®, Gallium Standard Solution, 10000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Supplier: VWR Chemicals
VWRC455392GEA 488 SGD
VWRC455392G
VWR® ARISTAR®, Gallium Standard Solution, 10000 mg/L Ga in dilute Nitric Acid (from Ga), Standard for ICP
Standards Cationic Standards Gallium Standards
Ga in HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.