Hydrochloric acid 5%, ARISTAR® for calibration blank in ICP

Supplier: VWR Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC456494PEA 196 SGD
VWRC456494P
Hydrochloric acid 5%, ARISTAR® for calibration blank in ICP
Hydrochloric acid

5% HCl in ASTM Type II Water


Ordering information: Supplied with certificate of analysis.

Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR