VWR® ARISTAR® Anion Multi-Component Standard 2 for Ion Chromatography, Avantor
Supplier: VWR Chemicals
VWRC458152EEA 748 SGD
VWRC458152E
VWR® ARISTAR® Anion Multi-Component Standard 2 for Ion Chromatography, Avantor
Standards Anionic Standards Multi-Element Anioinic Standards
Br 400 ppm   NO3 400 ppm
Cl 200 ppm   PO4 600 ppm
F 100 ppm   SO4 400 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.