Hydrochloric acid 36% VLSI, Electronic Grade

Supplier: VWR Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC50489136.320EA 415 SGD
VWRC50489136.320
Hydrochloric acid 36% VLSI, Electronic Grade
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
REACH: 01-2119484862-27

Order Now

Specification Test Results

Assay (acidimetric) 36.0 - 38.0 %
Colouration ≤ 10 APHA
Particle size(>0.50 μm)(/ml)(at filling) ≤ 64
Free halogen (as Cl₂) ≤ 0.5 ppm
Non-volatile matter ≤ 5
NH₄ (Ammonium) ≤ 1000 ppb
PO₄ (Phosphate) ≤ 100 ppb
SO₃ (Sulphite) ≤ 500 ppb
SO₄ (Sulphate) ≤ 500 ppb
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 50 ppb
As (Arsenic) ≤ 5 ppb
Au (Gold) ≤ 20 ppb
B (Boron) ≤ 10 ppb
Ba (Barium) ≤ 10 ppb
Be (Beryllium) ≤ 10 ppb
Bi (Bismuth) ≤ 20 ppb
Ca (Calcium) ≤ 200 ppb
Cd (Cadmium) ≤ 5 ppb
Co (Cobalt) ≤ 10 ppb
Cr (Chromium) ≤ 10 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 100 ppb
Ga (Gallium) ≤ 20 ppb
Ge (Germanium) ≤ 50 ppb
In (Indium) ≤ 10 ppb
K (Potassium) ≤ 50 ppb
Li (Lithium) ≤ 10 ppb
Mg (Magnesium) ≤ 50 ppb
Mn (Manganese) ≤ 10 ppb
Mo (Molybdenum) ≤ 10 ppb
Na (Sodium) ≤ 50 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Pt (Platinum) ≤ 20 ppb
Sn (Tin) ≤ 20 ppb
Sr (Strontium) ≤ 10 ppb
Ti (Titanium) ≤ 10 ppb
Tl (Thallium) ≤ 20 ppb
V (Vanadium) ≤ 10 ppb
Zn (Zinc) ≤ 50 ppb
Zr (Zirconium) ≤ 10 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR