BROXO salt, granules 6 - 15 mm

Supplier: VWR Chemicals

VWRC5254.9025EA 58 SGD
VWRC5254.9025
BROXO salt, granules 6 - 15 mm
Sodium chloride

Broxo water softener salt allows for problem free water softening that gives a great performance and is efficient in use. It is a very pure high-pressure compacted vacuum salt that dissolves evenly and gradually and is applicable to domestic and industrial installations. Broxo water softener can be used for both domestic and industrial use. The granules dissolves completely and evenly to leave no residual substances. The granules measure from 6mm up to 15mm and provide excellent performance. Also known as rock salt.


  • Very pure high-pressure compacted vacuum salt (99,8% NaCl)
  • For use in domestic and industrial installations
  • Granules from 6 mm up to 15 mm

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2,16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR