Orthoboric acid 10 g/l in aqueous solution, VOLUSOL®, with indicator

Supplier: VWR Chemicals

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

VWRC5732.5000EA 55.2 SGD
VWRC5732.5000
Orthoboric acid 10 g/l in aqueous solution, VOLUSOL®, with indicator
ortho-Boric acid
Formula: B(OH)₃
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (W/V) 0.9 - 1.1 %
Appearance Clear green liquid
Sensitivity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR