VWR® Emergency Intervention Kit
Supplier: VWR Chemicals
VWRC5791.0001EA 1160 SGD
VWRC5791.0001
VWR® Emergency Intervention Kit
Spill Control Materials
Set for absorption of organics, aqueous solutions, alkalis and acids. It contains.
  • 3× MERCK Chemizorb for absorption of organics and aqueous solutions (1.02051.0500)
  • MERCK Chemizorb for absorption of alkalis (including a neutraliser and a pH indicator) (1.01596.1000)
  • Absorbent PIG absorb­and­lock acid PLP501
  • Pair of barrier gloves with liner size 9 (112-1010)
  • Pair of barrier gloves with liner size 7 (112-0946)
  • 2× masks 4279 3M ABEK (111-0044)
  • 2× goggles 3M 2890A (111-0434)
  • Trash tray ergonomic (129-0174)
  • Brush in PBT, sterilisable (129-0175)
  • Disposal bag 48
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable