Sodium chloride 9 g/l in aqueous solution

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC5929.1000EA 21.5 SGD
VWRC5929.1000
Sodium chloride 9 g/l in aqueous solution
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Assay (W/V) 0.891 to 0.909 %
Appearance Clear colourless liquid
ZRAW Passes test Ph. Eur.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR