VWR® Dehydrated Culture Media in Jars
Supplier: BALIS
6001.00900 6001.01125
BALI6001.00900EA 162 SGD
BALI6001.00900 BALI6001.01125 VWRC6012.05000
VWR® Dehydrated Culture Media in Jars
Media for Microbiology Prepared Media for Microbiology
For the preliminary, non selective enrichment of bacteria, particularly pathogenic enterobacteriaceae, from foodstuffs.
  • Convenient, just add water
  • Delivered in jars