Sodium chloride 26% in aqueous solution, VOLUSOL®

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC6241.9011EA 331 SGD
VWRC6241.9011
Sodium chloride 26% in aqueous solution, VOLUSOL®
Sodium chloride
Formula: NaCl
Density: 1,195 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR