VWR® Silicone Fluids
Supplier: VWR Chemicals
VWRC630057WEA 9170 SGD
VWRC630057W VWRC630076C VWRC630077D VWRC630094E VWRC630096G VWRC630097H
VWR® Silicone Fluids
Circulator Bath Fluids
A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.
  • Non corrosive
  • High flash point
  • Long service life
  • Low evaporation
Fluids possess excellent heat transfer characteristics and cover a large temperature range from −40 to +200 °C.
1 cS = 1 mm²/s
Packaging: Packed in a plastic bottle/plastic container.
Caution: Harmful vapours may be given off at elevated temperatures. It is advisable to use this material in a fume cupboard at temperatures greater than 140 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable