VWR® MediaBag TW Dehydrated Culture Media in Bags
Supplier: VWR Chemicals
VWRC7101.00450EA 1150 SGD
VWRC7101.00450
VWR® MediaBag TW Dehydrated Culture Media in Bags
Media for Microbiology Prepared Media for Microbiology
MediaBags TW (Twirl-bag) are filled with pre-weighed granulated media, double-bagged bag and irradiated. Add sterile water under aseptic conditions.