VWR®, Taq DNA polymerase, 200000 U
Supplier: VWR Chemicals
VWRC733-2479EA 50730 SGD
VWRC733-2479
VWR®, Taq DNA polymerase, 200000 U
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq DNA polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from thermus aquaticus and has a molecular weight of approximately 94 kDa.
  • Store at –20 °C
Enzyme is supplied in 20 mM tris-HCl pH 8,5, 100 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5% tween 20, 0,5% NP40, 50% glycerol.
Caution: Reagent for in vitro laboratory use only.