Water, PCR grade

Supplier: VWR Chemicals

VWRC733-2573EA 111 SGD
VWRC733-2573
Water, PCR grade
Water

Ultrapure water, free of endonuclease-, nicking-, and exonuclease activity, free of human DNA.


  • Convenient 5 ml aliquots
  • Tested for contaminating activities


Caution: Storage at room temperature. Long term storage at -20 °C. Product expiry at -20 °C is stated on the label.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Freezer
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

DNA nicking activity Passed
Endonuclease activity Passed
Exonuclease activity Passed
Absence of Human DNA Passed
Volume check, 5 ml/tube Passed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR