VWR® Gurr™ Histological Fixatives, AFA
Supplier: VWR Chemicals
VWRC81009.362EA 106 SGD
VWRC81009.362
VWR® Gurr™ Histological Fixatives, AFA
Stains Microscopy and staining reagents
Acidified alcoholic formalin for the fixation of glycogen.
Contains acetic acid, ethanol and formaldehyde.