VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
VWRC83605.260EA 21.8 SGD
VWRC83605.260 VWRC83605.290 VWRC83606.260 VWRC83606.290 VWRC83605.180
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Electrolytes Electrode Filling Solutions
Potassium chloride 3 Molar solution may also be used as an electrode storage solution.
For full information concerning health and safety data please see the SDS on the website.