VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
VWRC83605.260EA 21.8 SGD
VWRC83605.260 VWRC83605.290 VWRC83606.260 VWRC83606.290 VWRC83605.180
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Electrolytes Electrode Filling Solutions
Potassium chloride 3 Molar solution may also be used as an electrode storage solution.
For full information concerning health and safety data please see the SDS on the website.
Order Now
SPECIFICATIONS
Environmentally Preferable