Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83622.320EA 207 SGD
VWRC83622.320
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Iso-Hexane
Formula: C₆H₁₄
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: –154 °C
Density: 0,653 g/cm³ (25 °C)
Flash Pt: –33 °C
Storage Temperature: Ambient
CAS Number: 64742-49-0
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce61b1646b637970765dc4b16a997650aeccfd1&vid=71606860b34952f73fd29b7df735dd0916b1e93c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless liquid
Assay (Mix. of C₆H₁₄ isomers) Min. 97.00 %
Water (KF) Max. 0.0100 % w/w
Residue after evaporation Max. 0.0005 % w/w
n-Hexane Max. 3.0 %
Transmittance (205 nm) Min. 50.0 %
Transmittance (220 nm) Min. 80.0 %
Transmittance (230 nm) Min. 95.0 %
Transmittance (280 nm) Min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR