Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83625.320EA 86.6 SGD
VWRC83625.320 VWRC83625.290
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Toluene
Formula: C₆H₅CH₃
Boiling Pt: 110,6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0,867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99,8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.05 %
Absorbance (300 nm) Max. 0.15
Absorbance (310 nm) Max. 0.1
Absorbance (330 nm) Max. 0.02
Absorbance (350 nm) Max. 0.01
Transmittance (300 nm) Min. 70 %
Transmittance (310 nm) Min. 80 %
Transmittance (330 nm) Min. 95 %
Transmittance (350 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR