1,4-Dioxane ≥99.8% unstabilised, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83628.320EA 519 SGD
VWRC83628.320
1,4-Dioxane ≥99.8% unstabilised, HiPerSolv CHROMANORM® for HPLC
1,4-Dioxane
Formula: C₄H₈O₂
Boiling Pt: 101,1 °C (1013 hPa)
Melting Pt: 11,8 °C
Density: 1,03 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.80 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 0.0005 %
Water Max. 0.0500 %
Transmittance (245 nm) Min. 50.0 %
Transmittance (270 nm) Min. 80.0 %
Transmittance (300 nm) Min. 98.0 %
Conforms to BDH 15290 Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR