1-Butanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83633.290EA 161 SGD
VWRC83633.290
1-Butanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
Boiling Pt: 117,6 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0,812 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.8 %
Acidity (as CH3COOH) Max. 0.002 %
Colour Max. 10 APHA
Residue after evaporation Max. 0.0005 %(m)
Water (KF) Max. 0.05 %(m)
Transmittance (210 nm) Min. 20 %
Transmittance (230 nm) Min. 70 %
Transmittance (270 nm) Min. 90 %
Transmittance (310 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR