N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83634.320EA 89.3 SGD
VWRC83634.320
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –60,5 °C
Density: 0,949 g/cm³ (20 °C)
Flash Pt: 57,5 °C (DIN 51755) (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.02 %
Transmittance (270 nm) Min. 10 %
Transmittance (275 nm) Min. 50 %
Transmittance (290 nm) Min. 80 %
Transmittance (300 nm) Min. 90 %
Transmittance (330 nm) Min. 98 %
Fluorescence (as quinine) (365 nm) Max. 1 ppb
Conforms to BDH 15297 Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR