N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83636.350EA 334 SGD
VWRC83636.350
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
Boiling Pt: 166,1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0,937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
REACH: 01-2119459339-27
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.50 %
Evaporation residue Max. 0.0005 %
Water Max. 500 ppm
Transmittance (270 nm) Min. 10.0 %
Transmittance (290 nm) Min. 98.0 %
Transmittance (310 nm) Min. 89.00 %
Transmittance (320 nm) Min. 94.0 %
Transmittance (400 nm) Min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR