Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83645.320EA 30 SGD
VWRC83645.320 VWRC83645.290 VWRC83645.400
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1,000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Colouration Max. 5 APHA
Conductivity (25°C) (at filling) Max. 1 µS/cm
Evaporation residue Max. 1 ppm
Al (Aluminium) Max. 50 ppb
Ca (Calcium) Max. 50 ppb
Fe (Iron) Max. 50 ppb
K (Potassium) Max. 50 ppb
Mg (Magnesium) Max. 50 ppb
Na (Sodium) Max. 100 ppb
Fluorescence (as quinine) (254/450 nm) Max. 1 ppb
Fluorescence (as quinine) (365/450 nm) Max. 0.5 ppb
Gradient grade (210 nm) Max. 5 mAU
Gradient (220 nm) Max. 2 mAU
Gradient (254 nm) Max. 1 mAU
MS-ESI+ (as Reserpine) Max. 2 ppb
MS-APCI+ (as Reserpine) Max. 2 ppb
MS-ESI- (as 4-Nitrophenol) Max. 20 ppb
MS-APCI- (as 4-Nitrophenol) Max. 20 ppb
Colony count (Ph.Eur.) (at filling) Max. 100 CFU/ml
TOC (at filling) Max. 30 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR