Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Filtered through a 0.1 micron filter.

VWRC83647.290EA 531 SGD
VWRC83647.290
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071

Order Now

Specification Test Results

Assay (GC) min. 99.9%
Identity (IR) complying
Clear colorless liquid complying
Color (APHA Pt-Co) max. 10
Water(KF) max. 0.01% w/w
Residue after evaporation max. 0.7 ppm
Acidity max 0.001 meq/g
Alkalinity max. 0.0002 meq/g
Aluminium (Al) max. 5 ppb
Calcium (Ca) max. 5 ppb
Iron (Fe) max. 5 ppb
Potassium (K) max. 5 ppb
Magnesium (Mg) max. 5 ppb
Sodium (Na) max. 25 ppb
Fluorescence 254/450 nm (as Quinine) max. 0.3 ppb
Fluorescence 365/450 nm (as Quinine) max. 0.3 ppb
UHPLC/MS ESI/APCI+ (as Reserpine) max. 2 ppb
UHPLC ESI/APCI- (as Reserpine) max. 10 ppb
PEG Signal-to-noise ratio max. 50
UHPLC-UV Gradient (210 nm) max. 1.0 mAU
UHPLC-UV Gradient (254 nm) max. 0.2 mAU
UV Absorbance 190 nm max. 1.00 AU
UV Absorbance 195 nm max. 0.15 AU
UV Absorbance 200 nm max. 0.05 AU
UV Absorbance 205 nm max. 0.04 AU
UV Absorbance 210 nm max. 0.03 AU
UV Absorbance 215 nm max. 0.025 AU
UV Absorbance 220 nm max. 0.015 AU
UV Absorbance 225 nm max. 0.015 AU
UV Absorbance 230 nm max. 0.010 AU
UV Absorbance 240 nm max. 0.005 AU
UV Absorbance 254-400 nm max. 0.005 AU
Filtered through 0.1 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR