Sodium diethyldithiocarbamate trihydrate 97 zA

Supplier: VWR Chemicals
Warning

Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

VWRC83864.180EA 213 SGD
VWRC83864.180
Sodium diethyldithiocarbamate trihydrate 97 zA
Sodium diethyldithiocarbamate trihydrate
Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 95…98,5 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9 III
Merck Index: 12,03443

Order Now

Specification Test Results

Assay Min. 97 %
Ignition residue (SO4) 30.0 - 35.0 %
Suited for copper reagent Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR