Perchloric acid 65%, NORMATOM® for trace metal analysis

Supplier: VWR Chemicals
Danger

VWRC83874.270EA 420 SGD
VWRC83874.270
Perchloric acid 65%, NORMATOM® for trace metal analysis
Perchloric acid
Formula: HClO₄
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112,2 °C
Density: 1,53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 (8),I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 65 - 71 %
Colouration Max. 10 APHA
Free chlorine Max. 0.5 ppm
Ignition residue (SO₄) Max. 30 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 3 ppm
ClO₃ (Chlorate) Max. 10 ppm
PO₄+ SiO₄(as SiO₂) Max. 5 ppm
SO4 (Sulphate) Max. 10 ppm
Ag (Silver) Max. 1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsenic) Max. 0.5 ppb
Au (Gold) Max. 0.5 ppb
Ba (Barium) Max. 1 ppb
Be (Beryllium) Max. 0.5 ppb
Bi (Bismuth) Max. 0.5 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 1 ppb
Ce (Cerium) Max. 0.5 ppb
Co (Cobalt) Max. 0.5 ppb
Cs (Cesium) Max. 0.5 ppb
Cu (Copper) Max. 0.5 ppb
Dy (Dysprosium) Max. 0.5 ppb
Er (Erbium) Max. 0.5 ppb
Eu (Europium) Max. 0.5 ppb
Fe (Iron) Max. 1 ppb
Ga (Gallium) Max. 0.5 ppb
Gd (Gadolinium) Max. 0.5 ppb
Ho (Holmium) Max. 0.5 ppb
In (Indium) Max. 0.5 ppb
K (Potassium) Max. 1 ppb
La (Lanthanum) Max. 0.5 ppb
Li (Lithium) Max. 0.5 ppb
Lu (Lutetium) Max. 0.5 ppb
Mg (Magnesium) Max. 1 ppb
Mn (Manganese) Max. 1 ppb
Mo (Molybdenum) Max. 0.5 ppb
Na (Sodium) Max. 1 ppb
Nd (Neodymium) Max. 0.5 ppb
Ni (Nickel) Max. 1 ppb
Pb (Lead) Max. 1 ppb
Pd (Palladium) Max. 0.5 ppb
Pr (Praseodymium) Max. 0.5 ppb
Pt (Platinum) Max. 0.5 ppb
Rb (Rubidium) Max. 0.5 ppb
Rh (Rhodium) Max. 0.5 ppb
Ru (Ruthenium) Max. 0.5 ppb
Sb (Antimony) Max. 0.5 ppb
Sc (Scandium) Max. 0.5 ppb
Sm (Samarium) Max. 0.5 ppb
Sn (Tin) Max. 1 ppb
Sr (Strontium) Max. 0.5 ppb
Tb (Terbium) Max. 0.5 ppb
Te (Tellurium) Max. 0.5 ppb
Th (Thorium) Max. 1 ppb
Ti (Titanium) Max. 1 ppb
Tl (Thallium) Max. 0.5 ppb
Tm (Thulium) Max. 0.5 ppb
U (Uranium) Max. 0.5 ppb
V (Vanadium) Max. 0.5 ppb
Y (Yttrium) Max. 0.5 ppb
Yb (Ytterbium) Max. 0.5 ppb
Zn (Zinc) Max. 1 ppb
Zr (Zirconium) Max. 0.5 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR