Water, NORMATOM® for trace metal analysis

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

VWRC83877.260EA 109 SGD
VWRC83877.260 VWRC83877.290
Water, NORMATOM® for trace metal analysis
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Colouration 10 CO max.
Ag (Silver) Max. 50 ppt
Al (Aluminium) Max. 200 ppt
As (Arsenic) Max. 50 ppt
Au (Gold) Max. 50 ppt
B (Boron) Max. 500 ppt
Ba (Barium) Max. 50 ppt
Be (Beryllium) Max. 50 ppt
Bi (Bismuth) Max. 50 ppt
Ca (Calcium) Max. 500 ppt
Cd (Cadmium) Max. 50 ppt
Ce (Cerium) Max. 20 ppt
Co (Cobalt) Max. 50 ppt
Cr (Chromium) Max. 50 ppt
Cs (Cesium) Max. 50 ppt
Cu (Copper) Max. 100 ppt
Dy (Dysprosium) Min. 20 ppt
Er (Erbium) Max. 20 ppt
Eu (Europium) Max. 20 ppt
Fe (Iron) Max. 300 ppt
Ga (Gallium) Max. 50 ppt
Gd (Gadolinium) Max. 20 ppt
Ge (Germanium) Max. 50 ppt
Hf (Hafnium) Max. 10 ppt
Hg (Mercury) Max. 200 ppt
Ho (Holmium) Max. 20 ppt
In (Indium) Max. 50 ppt
K (Potassium) Max. 300 ppt
La (Lanthanum) Max. 20 ppt
Li (Lithium) Max. 50 ppt
Lu (Lutetium) Max. 20 ppt
Mg (Magnesium) Max. 100 ppt
Mn (Manganese) Max. 50 ppt
Mo (Molybdenum) Max. 50 ppt
Na (Sodium) Max. 500 ppt
Nb (Niobium) Max. 20 ppt
Nd (Neodymium) Max. 20 ppt
Ni (Nickel) Max. 50 ppt
Pb (Lead) Max. 50 ppt
Pd (Palladium) Max. 50 ppt
Pr (Praseodymium) Max. 20 ppt
Pt (Platinum) Max. 50 ppt
Rb (Rubidium) Max. 20 ppt
Re (Rhenium) Max. 50 ppt
Rh (Rhodium) Max. 50 ppt
Ru (Ruthenium) Max. 50 ppt
Sb (Antimony) Max. 50 ppt
Sc (Scandium) Max. 20 ppt
Se (Selenium) Max. 200 ppt
Sm (Samarium) Max. 20 ppt
Sn (Tin) Max. 50 ppt
Sr (Strontium) Max. 50 ppt
Ta (Tantalum) Max. 50 ppt
Tb (Terbium) Max. 20 ppt
Te (Tellurio) Max. 50 ppt
Th (Thorium) Max. 10 ppt
Ti (Titanium) Max. 50 ppt
Tl (Thallium) Max. 50 ppt
Tm (Thulium) Max. 20 ppt
U (Uranium) Max. 10 ppt
V (Vanadium) Max. 50 ppt
W (Tungsten) Max. 100 ppt
Y (Yttrium) Max. 10 ppt
Yb (Ytterbium) Max. 20 ppt
Zn (Zinc) Max. 500 ppt
Zr (Zirconium) Max. 50 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR