Water, NORMATOM®, ultrapure Trace_metal

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

VWRC83880.290EA 637 SGD
VWRC83880.290
Water, NORMATOM®, ultrapure Trace_metal
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Colouration (APHA) max. 10
Cl (Chloride) max. 1 ppb
PO4 (Phosphate) max. 1 ppb
SO4 (Sulphate) max. 1 ppb
Ag (Silver) max. 10 ppt
Al (Aluminium) max. 20 ppt
As (Arsenic) max. 10 ppt
Au (Gold) max. 10 ppt
B (Boron) max. 20 ppt
Ba (Barium) max. 10 ppt
Be (Beryllium) max. 10 ppt
Bi (Bismuth) max. 10 ppt
Ca (Calcium) max. 10 ppt
Cd (Cadmium) max. 10 ppt
Ce (Cerium) max. 10 ppt
Co (Cobalt) max. 10 ppt
Cr (Chromium) max. 10 ppt
Cs (Cesium) max. 10 ppt
Cu (Copper) max. 10 ppt
Dy (Dysprosium) max. 1 ppt
Er (erbium) max. 1 ppt
Eu (Europium) max. 1 ppt
Fe (Iron) max. 10 ppt
Ga (Gallium) max. 10 ppt
Gd (Gadolinium) max. 1 ppt
Ge (Germanium) max. 10 ppt
Hf (Hafnium) max. 1 ppt
Hg (Mercury) max. 20 ppt
Ho (Holmium) max. 1 ppt
In (Indium) max. 1 ppt
K (Potassium) max. 10 ppt
La (Lanthanum) max. 1 ppt
Li (Lithium) max. 10 ppt
Lu (Lutetium) max. 1 ppt
Mg (Magnesium) max. 10 ppt
Mn (Manganese) Max. 20 ppt
Mo (Molybdenum) max. 10 ppt
Na (Sodium) max. 10 ppt
Nb (Niobium) max. 10 ppt
Nd (Neodymium) max. 1 ppt
Ni (Nickel) max. 10 ppt
Pb (Lead) max. 10 ppt
Pd (Palladium) max. 10 ppt
Pr (Praseodymium) max. 10 ppt
Pt (Platinum) max. 10 ppt
Rb (Rubidium) max. 10 ppt
Re (Rhenium) max. 10 ppt
Rh (Rhodium) max. 10 ppt
Ru (Ruthenium) max. 10 ppt
Sb (Antimony) max. 10 ppt
Sc (Scandium) max. 10 ppt
Se (Selenium) max. 50 ppt
Sm (Samarium) max. 10 ppt
Sn (Tin) max. 10 ppt
Sr (Strontium) max. 10 ppt
Ta (Tantalum) max. 10 ppt
Tb (Terbium) max. 10 ppt
Te (Tellurium) max. 1 ppt
Th (Thorium) max. 1 ppt
Ti (Titanium) max. 10 ppt
Tl (Thallium) max. 10 ppt
Tm (Thulium) max. 10 ppt
U (Uranium) max. 1 ppt
V (Vanadium) max. 10 ppt
W (Tungsten) max. 10 ppt
Y (Yttrium) max. 1 ppt
Yb (Ytterbium) max. 10 ppt
Zn (Zinc) max. 10 ppt
Zr (Zirconium) max. 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR