n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83962.320EA 262 SGD
VWRC83962.320
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.0 %
Acidity Max. 0.0005 meq/g
Evaporation residue (100°C) Max. 0.0005 %
Water Max. 0.01 %
Organic residue (as Octanol) Max. 10 ng/ml
Halogenated residue (as Lindane)(GC/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR