Ethyl acetate ≥99.8%, PESTINORM® for capillary GC analysis

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid ethyl ester

Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83963.320EA 119 SGD
VWRC83963.320
Ethyl acetate ≥99.8%, PESTINORM® for capillary GC analysis
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue (100°C) Max. 0.0005 %
Water Max. 0.05 %
Organic residue (as Octanol) (GC/FID) Max. 10 ng/l
Halogenated residue (as Lindane)(GC/ECD) Max. 5 ng/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR