Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Hexanes, Isohexane

Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC83992.320EA 87 SGD
VWRC83992.320
Hexane (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 45.0 %
Acidity Max. 0.001 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.01 %
Transmittance (210 nm) Min. 60 %
Transmittance (220 nm) Min. 80 %
Transmittance (230 nm) Min. 95 %
Transmittance (240 nm) Min. 98 %
Transmittance (250 nm) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR