Sulphuric acid 62% for the determination of fats in cheese according to Gerber

Supplier: VWR Chemicals
Danger

VWRC84509.290EA 55.4 SGD
VWRC84509.290 VWRC84509.360
Sulphuric acid 62% for the determination of fats in cheese according to Gerber
Sulphuric acid
Formula: H₂SO₄
Boiling Pt: >100 °C (1013 hPa)
Density: 1,5…1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Density (20/4) 1.517 - 1.527
Assay 61.7 - 62.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR