Silicone antifoam 15% (dry compound) for use in aqueous, textile, chemical and industrial foam control

Supplier: VWR Chemicals

VWRC84537.290EA 115 SGD
VWRC84537.290 VWRC84537.230
Silicone antifoam 15% (dry compound) for use in aqueous, textile, chemical and industrial foam control
Silicone antifoam

Viscosity (for information only): 600 to 1.500 mPA.s

Order Now

Specification Test Results

Dry matter content 14.0 - 17.0 %
Density (25/4) 0.950 - 1.100
pH (20°C) 4.5 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR