Silicone oil, 500 cSt

Supplier: VWR Chemicals

VWRC84539.290EA 118 SGD
VWRC84539.290
Silicone oil, 500 cSt
Silicone oil
Density: 0,97 g/cm³ (20 °C)
Flash Pt: 321 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00132673
CAS Number: 68083-14-7

Order Now

Specification Test Results

Viscosity (25°C) 450 - 550 cSt
Volatile matter Max. 1.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR